Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABARAP Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP171771Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171771Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171771Z Supplier Novus Biologicals Supplier No. NBP171771AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABARAP Polyclonal specifically detects GABARAP in Human, Mouse, Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen GABARAP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O95166
Gene Alias ATG8A, FLC3B, FLJ25768, GABA(A) receptor-associated proteinMGC120154, gamma-aminobutyric acid receptor-associated protein, MM46MGC120155
Gene Symbols GABARAP
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the human GABARAP protein (within residues 50-117). [Swiss-Prot O95166]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Golgi Apparatus Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 11337
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Target Species Human, Mouse, Drosophila
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.