Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABBR1 Monoclonal Antibody (N93A/49), APC
GREENER_CHOICE

Catalog No. PIMA545536
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545536 Supplier Invitrogen™ Supplier No. MA545536
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

1 μg/mL of MA5-45536 was sufficient for detection of GABA(B)R1 in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 115kDa. No cross-reactivity against GABA(B)R2. This antibody was formerly sold as clone S93A-49.

Receptor for GABA. The activity of this receptor is mediated by G-proteins that inhibit adenylyl cyclase activity, stimulates phospholipase A2, activates potassium channels, inactivates voltage-dependent calcium-channels and modulates inositol phospholipids hydrolysis. Plays a critical role in the fine-tuning of inhibitory synaptic transmission. Pre-synaptic GABA-B-R inhibit neurotransmitter release by down-regulating high-voltage activated calcium channels, whereas postsynaptic GABA-B-R decrease neuronal excitability by activating a prominent inwardly rectifying potassium (Kir) conductance that underlies the late inhibitory postsynaptic potentials. Not only implicated in synaptic inhibition but also in hippocampal long-term potentiation, slow wave sleep, muscle relaxation and antinociception.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABBR1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N93A/49
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene GABBR1
Gene Accession No. Q9UBS5, Q9WV18, Q9Z0U4
Gene Alias bM573K1.1; bM573K1.1.1 (gamma-aminobutyric acid (GABA) B receptor, 1a); DAAP-188P13.3; dJ271M21.1.1; dJ271M21.1.2; FLJ92613; GABA(B) receptor; GABAB; GABA-B receptor 1; GABA-B receptor, R1 subunit; GABAB1; Gaba-b1; GABAbR1; GABA-BR1; GABA-B-R1; Gababr1 receptor; Gaba-b-r1a; Gaba-br1a receptor; GABA-BR1b receptor; Gabbr1; GABBR1-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) B receptor 1; gamma-aminobutyric acid (GABA) B receptor, 1; gamma-aminobutyric acid (GABA-B) receptor, 1; gamma-aminobutyric acid type B receptor subunit 1; GB1; GBR1; GPRC3A; HGB1a; OTTHUMP00000109099; OTTHUMP00000214725; seven transmembrane helix receptor
Gene Symbols GABBR1
Host Species Mouse
Immunogen Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2550, 54393, 81657
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.