Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Antibody (GAD1/2391), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB387628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387628 Supplier Novus Biologicals Supplier No. NBP308276JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen GAD1/GAD67
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone GAD1/2391
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Host Species Mouse
Immunogen Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) (Uniprot: Q99259)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2571
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.