Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106989 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106989 Supplier Novus Biologicals Supplier No. NBP106989
Add to Cart
Add to Cart

Goat Polyclonal Antibody

GAD1/GAD67 Polyclonal specifically detects GAD1/GAD67 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Peptide ELISA.

Specifications

Antigen GAD1/GAD67
Applications Western Blot, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1.5 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:2000, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q99259
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Gene Symbols GAD1
Host Species Goat
Immunogen Peptide with sequence C-PDSPQRREKLHK corresponding to internal region according to NP_000808.2.
Molecular Weight of Antigen 67 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2571
Test Specificity This antibody is expected to recognize isoform GAD67. There is no cross-reactivity expected with GAD2.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.