Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Antibody, Novus Biologicals™
SDP

Catalog No. NBP102161 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP102161 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP102161 Supplier Novus Biologicals Supplier No. NBP102161
Add to Cart
Add to Cart

Chicken Polyclonal Antibody has been used in 3 publications

GAD1/GAD67 Polyclonal specifically detects GAD1/GAD67 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GAD1/GAD67
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:5000, Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:1000-1:2000, Immunohistochemistry-Paraffin 1:1000-1:2000
Formulation 10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide
Gene Accession No. Q99259
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Gene Symbols GAD1
Host Species Chicken
Immunogen Chickens were immunized with a synthetic peptide/keyhole limpet hemocyanin (KLH) conjugate. This synthetic peptide corresponded to a region near the C-terminus of this gene product, and was 100% conserved between the human (Q99259, NCBI), mouse (P48318, NCBI) and rat (NP_058703, NCBI) gene products.
Molecular Weight of Antigen 67 kDa
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2571
Test Specificity GAD
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.