Learn More
Description
Specifications
Specifications
| Antigen | GAD1/GAD67 |
| Applications | Western Blot, Flow Cytometry, ELISA, Peptide Array |
| Classification | Monoclonal |
| Clone | GAD1/2563 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | Q99259, Q99259 |
| Gene Alias | 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
