Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67, Mouse anti-Human, Clone: GAD1/2563, Novus Biologicals™
SDP

Catalog No. NB010624 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010624 20 μg
NB010623 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB010624 Supplier Novus Biologicals Supplier No. NBP27980320UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Protein Array.

Specifications

Antigen GAD1/GAD67
Applications Western Blot, Flow Cytometry, ELISA, Peptide Array
Classification Monoclonal
Clone GAD1/2563
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q99259, Q99259
Gene Alias 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Gene Symbols GAD1
Host Species Mouse
Immunogen Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 2571
Test Specificity This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GAD1/GAD67).
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.