Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD2/GAD65 Antibody (GAD2/2362), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB379049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379049 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379049 Supplier Novus Biologicals Supplier No. NBP308704R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen GAD2/GAD65
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone GAD2/2362
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase
Host Species Mouse
Immunogen Recombinant fragment of human GAD2/GAD65 protein (around aa 6-99) (exact sequence is proprietary) (Uniprot: Q05329)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2572
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.