Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD153/CHOP Antibody (9C8), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB6001335R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001335R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001335R Supplier Novus Biologicals Supplier No. NB6001335R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GADD153/CHOP Monoclonal specifically detects GADD153/CHOP in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen GADD153/CHOP
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone 9C8
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Gene Accession No. P35638
Gene Alias C/EBP-homologous protein, C/EBP-homologous protein 10, CCAAT/enhancer-binding protein homologous protein, CHOP-10, CHOP10 CEBPZ, CHOPC/EBP zeta, DDIT-3, DNA damage-inducible transcript 3 protein, DNA-damage-inducible transcript 3, GADD153, Growth arrest and DNA damage-inducible protein GADD153
Gene Symbols DDIT3
Host Species Mouse
Immunogen Full length mouse CHOP/GADD153 [Swiss-Prot# P35639]
Molecular Weight of Antigen 29 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, DNA Repair, Hypoxia, Lipid and Metabolism, Neuroscience, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 1649
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.