Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Antibody (9E10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600302SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600302SS 0.025 mL
NB600302 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600302SS Supplier Novus Biologicals Supplier No. NB600302SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 33 publications

c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Bovine, Drosophila samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, Sandwich ELISA, Flow (Intracellular).

Specifications

Antigen c-Myc
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA
Classification Monoclonal
Clone 90000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2.0 ug/ml, Simple Western 1:200, Flow Cytometry 1:50-1:200. Use reported in scientific literature (PMID 21315712), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Immunoblotting reported in scientific literature (PMID 23750001), Proximity Ligation Assay reported in scientific literature (PMID 33298911), Sandwich ELISA reported in scientific literature (PMID 33122198), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP)
Gene Accession No. P01106
Gene Alias 9E10 anti-Myc, 9E10 Myc, avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
Gene Symbols MYC
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) of human c-Myc Antibody (9E10). [UniProt# P01106]
Molecular Weight of Antigen 48.8 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4609
Test Specificity Specific for the c-myc protein in random coil configuration, not as a helix. 9E10 does not react with V-myc.
Target Species Human, Mouse, Bovine, Drosophila
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.