Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Myc Antibody (9E10) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 33 publications

$206.50 - $512.00

Specifications

Antigen c-Myc
Clone 90000000000
Concentration 1 mg/mL
Dilution Western Blot 0.5-2.0 ug/ml, Simple Western 1:200, Flow Cytometry 1:50-1:200. Use reported in scientific literature (PMID 21315712), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Immunoblotting reported in scientific literature (PMID 23750001), Proximity Ligation Assay reported in scientific literature (PMID 33298911), Sandwich ELISA reported in scientific literature (PMID 33122198), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600302SS
SDP
View Documents
Novus Biologicals
NB600302SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB600302
SDP
View Documents
Novus Biologicals
NB600302
0.1 mL
Each for $512.00
Only null left
Add to cart
 
Description

Description

c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Bovine, Drosophila samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, Sandwich ELISA, Flow (Intracellular).
Specifications

Specifications

c-Myc
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In situ PLA
Unconjugated
Mouse
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
P01106
4609
A synthetic peptide corresponding to amino acids 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) of human c-Myc Antibody (9E10). [UniProt# P01106]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
48.8 kDa
90000000000
Western Blot 0.5-2.0 ug/ml, Simple Western 1:200, Flow Cytometry 1:50-1:200. Use reported in scientific literature (PMID 21315712), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen 1:50-1:200, Immunoblotting reported in scientific literature (PMID 23750001), Proximity Ligation Assay reported in scientific literature (PMID 33298911), Sandwich ELISA reported in scientific literature (PMID 33122198), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP)
Monoclonal
Purified
RUO
Human, Mouse, Bovine, Drosophila
9E10 anti-Myc, 9E10 Myc, avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)
MYC
IgG1 κ
Protein G purified
Specific for the c-myc protein in random coil configuration, not as a helix. 9E10 does not react with V-myc.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.