Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD34 Antibody (OTI3D12), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N271765J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271765J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271765J549 Supplier Novus Biologicals Supplier No. NBP271765JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GADD34 Monoclonal specifically detects GADD34 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen GADD34
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3D12
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias GADD34Growth arrest and DNA damage-inducible protein GADD34, growth arrest and DNA-damage-inducible 34, Myeloid differentiation primary response protein MyD116 homolog, protein phosphatase 1 regulatory subunit 15A, protein phosphatase 1, regulatory (inhibitor) subunit 15A
Gene Symbols PPP1R15A
Host Species Mouse
Immunogen Full length recombinant protein of human GADD34 (NP_055145) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Lipid and Metabolism, Unfolded Protein Response
Primary or Secondary Primary
Gene ID (Entrez) 23645.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.