Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD45 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | GADD45 alpha |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GADD45 alpha Polyclonal specifically detects GADD45 alpha in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GADD45 alpha | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
1647 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MTLEEFSAGEQKTERMDKVGDALEEVLSKALSQ | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha | |
GADD45A | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title