Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45 alpha Antibody (OTI1C9), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB289521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289521 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289521 Supplier Novus Biologicals Supplier No. NBP270559APC

Mouse Monoclonal Antibody

GADD45 alpha Monoclonal antibody specifically detects GADD45 alpha in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen GADD45 alpha
Applications Western Blot
Classification Monoclonal
Clone OTI1C9
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha
Host Species Mouse
Immunogen Full length human recombinant protein of human GADD45A(NP_001915) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1647
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.