Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45G Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB065312 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065312 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065312 Supplier Novus Biologicals Supplier No. NBP300285AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GADD45G Polyclonal specifically detects GADD45G in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GADD45G
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CR6DNA damage-inducible transcript 2 protein, Cytokine-responsive protein CR6, DDIT-2, DDIT2GADD45-gamma, growth arrest and DNA damage-inducible protein GADD45 gamma, growth arrest and DNA-damage-inducible, gamma, GRP1717 kD
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant GADD45G (O95257; Met1-Glu159).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10912
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.