Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gAdiponectin/gAcrp30 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB121191 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121191 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121191 Supplier Novus Biologicals Supplier No. NBP311025C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

gAdiponectin/gAcrp30 Polyclonal antibody specifically detects gAdiponectin/gAcrp30 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen gAdiponectin/gAcrp30
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Recombinant Human gAdiponectin/gAcrp30
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9370
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.