Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) carbonate hydrate, 99% (REO)

Catalog No. p-7051013
Click to view available options
Quantity:
5 g
25 g

CAS: 38245-36-2 | C3Gd2O9 | 494.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38245-36-2
Molecular Formula C3Gd2O9
Molecular Weight (g/mol) 494.52
MDL Number MFCD00150240
InChI Key RQXZRSYWGRRGCD-UHFFFAOYSA-H
Synonym gadolinium carbonate hydrate, gadolinium iii carbonate hydrate, acmc-1aeff, 3co3.2gd.h2o, gadolinium carbonate-water 2/3/1, digadolinium 3+ hydrate tricarbonate, gadolinium iii carbonate hydrate, reacton?, gadolinium iii carbonate hydrate-gd reo puratrem
PubChem CID 16212943
SMILES [Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Physical Form Powder
Quantity 5 g
IUPAC Name gadolinium(3+);tricarbonate;hydrate
Formula Weight 494.53 (Anhydrous)
Percent Purity 99%
Assay (REO)
Chemical Name or Material Gadolinium(III) carbonate hydrate
EINECSNumber 227-583-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.