Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Catalog Number AA1320714
Click to view available options
Quantity:
25 g
100 g
38245-36-2
C3Gd2O9
494.52
MFCD00150240
RQXZRSYWGRRGCD-UHFFFAOYSA-H
gadolinium carbonate hydrate, gadolinium iii carbonate hydrate, acmc-1aeff, 3co3.2gd.h2o, gadolinium carbonate-water 2/3/1, digadolinium 3+ hydrate tricarbonate, gadolinium iii carbonate hydrate, reacton?, gadolinium iii carbonate hydrate-gd reo puratrem
16212943
digadolinium(3+) tricarbonate
[Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
38245-36-2
C3Gd2O9
494.52
MFCD00150240
RQXZRSYWGRRGCD-UHFFFAOYSA-H
gadolinium carbonate hydrate, gadolinium iii carbonate hydrate, acmc-1aeff, 3co3.2gd.h2o, gadolinium carbonate-water 2/3/1, digadolinium 3+ hydrate tricarbonate, gadolinium iii carbonate hydrate, reacton?, gadolinium iii carbonate hydrate-gd reo puratrem
16212943
digadolinium(3+) tricarbonate
[Gd+3].[Gd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Gadolinium(III) carbonate is used for making optical glass and dopant for Gadolinium Yttrium Garnets which have microwave applications. It is also used for making phosphors for color TV tube and energy-saving lamps. It is used as intravenous MRI contrast agent to enhance images in medical magnetic resonance imaging and Magnetic Resonance Angiography (MRA) procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 494.53 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Assay (REO)
Chemical Name or Material Gadolinium(III) carbonate hydrate
EINECSNumber 227-583-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit