Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAGE1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | GAGE1 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GAGE1 Polyclonal specifically detects GAGE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GAGE1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
2543 | |
This antibody was developed against a Recombinant Protein corresponding to amino acids:PPEMIGPMRPEQFSDEVEPATPEEGEPATQRQDPAAA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Polyclonal | |
Rabbit | |
Cancer | |
Antigen MZ2-F, Cancer/testis antigen 4.1, G antigen 1, GAGE-1, member 1, MGC33825, MZ2-F antigen | |
GAGE1 | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title