Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAK Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB289856 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289856 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289856 Supplier Novus Biologicals Supplier No. NBP232181PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GAK Polyclonal antibody specifically detects GAK in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen GAK
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias cyclin G associated kinase, cyclin-G-associated kinase, EC 2.7.11.1, FLJ16629, FLJ40395, MGC99654
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 675 to 725 of human Cyclin G Associated Kinase using the numbering given in entry NP_005246.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2580
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.