Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin Antibody (581403), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB303180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303180 Supplier Novus Biologicals Supplier No. FAB5854U

Mouse Monoclonal Antibody

Galanin Monoclonal antibody specifically detects Galanin in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF-reported

Specifications

Antigen Galanin
Applications Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 581403
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167
Host Species Mouse
Immunogen E.coli-derived recombinant human Galanin, Ala20-Ser123, Accession # P22466
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51083
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.