Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin Antibody, Novus Biologicals™
SDP

Catalog No. NBP145217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145217 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145217 Supplier Novus Biologicals Supplier No. NBP145217
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

Galanin Polyclonal specifically detects Galanin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Peptide ELISA.

Specifications

Antigen Galanin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry, Immunohistochemistry-Frozen 0.2 - 0.5 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P22466
Gene Alias GAL1, galanin, galanin prepropeptide, GALNgalanin-related peptide, GLNNgalanin-message-associated peptide, GMAP, MGC40167
Gene Symbols GAL
Host Species Goat
Immunogen Peptide with sequence C-HRSFSDKNGLTSK corresponding to internal region according to NP_057057.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 51083
Test Specificity This antibody recognizes and epitope within the internal region. Human, Mouse. Predicted cross-reactivity based on sequence identity: Rat, Cow.
Target Species Human, Mouse, Rat, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.