Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin R1/GALR1 Antibody (613207) [Alexa Fluor™ 532], Novus Biologicals™
SDP

Catalog No. NB401700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401700 Supplier Novus Biologicals Supplier No. FAB10353X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galanin R1/GALR1 Monoclonal antibody specifically detects Galanin R1/GALR1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Galanin R1/GALR1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 613207
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias galanin receptor 1, galanin receptor type 1, GALNRGALNR1GAL1-R, GALR-1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Galanin R1/GALR1, Accession # P47211
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2587
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.