Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galanin R2/GALR2 Antibody (616520), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346940 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346940 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346940 Supplier Novus Biologicals Supplier No. FAB6544MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galanin R2/GALR2 Monoclonal antibody specifically detects Galanin R2/GALR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF

Specifications

Antigen Galanin R2/GALR2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 616520
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias GAL2-R, galanin receptor 2, galanin receptor type 2, GALNR2MGC125984, GALR-2, MGC125983
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Galanin R2, Accession # O43603
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 8811
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.