Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALE Antibody (6G10), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB160941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160941 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160941 Supplier Novus Biologicals Supplier No. NBP259421FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

GALE Monoclonal antibody specifically detects GALE in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen GALE
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 6G10
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 5.1.3, EC 5.1.3.2, FLJ95174, FLJ97302, Galactowaldenase, short chain dehydrogenase/reductase family 1E, member 1, UDP-, UDP galactose-4'-epimerase, UDP-galactose 4-epimerase, UDP-galactose-4-epimerase, UDP-glucose 4-epimerase
Host Species Mouse
Immunogen Recombinant human GALE (1-348aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2582
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.