Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-1 Antibody (RM0081-9J26), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP12721646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP12721646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP12721646 Supplier Novus Biologicals Supplier No. NBP121721JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Galectin-1 Monoclonal specifically detects Galectin-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Galectin-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0081-9J26
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 14 kDa laminin-binding protein, 14 kDa lectin, Beta-galactoside-binding lectin L-14-I, DKFZp686E23103, gal-1, GAL1, Galaptin, galectin 1, galectin-1, GBP, HBL, HLBP14, HPL, Lactose-binding lectin 1, Lectin galactoside-binding soluble 1, lectin, galactoside-binding, soluble, 1, Putative MAPK-activating protein PM12, S-Lac lectin 1
Gene Symbols LGALS1
Host Species Rat
Immunogen Mouse recombinant Galectin-1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3956
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Galectin-1.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.