Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-10 Antibody (561603), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB367967 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367967 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367967 Supplier Novus Biologicals Supplier No. FAB5447Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin-10 Monoclonal specifically detects Galectin-10 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen Galectin-10
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 561603
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Charcot-Leyden crystal proteinLGALS10A, EC 3.1.1.5, eosinophil lysophospholipase, Gal-10, GAL10, galectin 10, galectin-10, LGALS10, LPPL_HUMAN, Lysolecithin acylhydrolase, MGC149659
Host Species Mouse
Immunogen E. coli-derived recombinant human Galectin-10, Met1-Arg142, Accession # Q05315
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1178
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.