Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin 13 Antibody (PP13/1164), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB008163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB008163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB008163 Supplier Novus Biologicals Supplier No. NBP254397G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin 13 Monoclonal specifically detects Galectin 13 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Galectin 13
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PP13/1164
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13
Gene Symbols LGALS13
Host Species Mouse
Immunogen Recombinant human Galectin 13 protein fragment (aa23-134) (Uniprot: Q9UHV8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.