Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin 13 Antibody (PP13/1164), Novus Biologicals™
SDP

Catalog No. NB253167100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253167100 100 μg
NB25316720 20 μg
NB253167200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253167100 Supplier Novus Biologicals Supplier No. NBP253167100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin 13 Monoclonal specifically detects Galectin 13 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Galectin 13
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PP13/1164
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 1mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias beta-galactoside-binding lectin, EC 3.1.1.5, gal-13, GAL13, galectin-13, lectin, galactoside-binding, soluble, 13, PLAC8galectin 13, Placental protein 13, Placental tissue protein 13, PP13galactoside-binding soluble lectin 13
Gene Symbols LGALS13
Host Species Mouse
Immunogen Recombinant human Galectin-13 protein fragment (aa23-134) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29124
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.