Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-2 Antibody (2509A), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB305193 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305193 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305193 Supplier Novus Biologicals Supplier No. FAB1153N

Rabbit Monoclonal Antibody

Galectin-2 Monoclonal antibody specifically detects Galectin-2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Galectin-2
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2509A
Conjugate Alexa Fluor 700
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Beta-galactoside-binding lectin L-14-II, Gal-2, galectin 2, galectin-2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, MGC75071, S-Lac lectin 2
Host Species Rabbit
Immunogen E. coli-derived human Galectin-2, Met1-Glu132, Accession # P05162
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3957
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.