Learn More
Description
Specifications
Specifications
| Antigen | Galectin-3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M3/38 |
| Conjugate | PE |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Gene Alias | Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3 |
| Gene Symbols | LGALS3 |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was Galectin3 Mac2. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
