Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3 Rabbit anti-Human, Clone: 24, Novus Biologicals™
SDP

Catalog No. NB020250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020250 50 μg
NB020249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020250 Supplier Novus Biologicals Supplier No. NBP28938350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Galectin-3 Monoclonal specifically detects Galectin-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Galectin-3
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 24
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 0.5-2ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Host Species Rabbit
Immunogen Recombinant Human Galectin-3 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3958
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.