Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271125B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271125B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271125B Supplier Novus Biologicals Supplier No. NBP271125B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin-3BP/MAC-2BP/LGALS3BP Monoclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Galectin-3BP/MAC-2BP/LGALS3BP
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI6B7
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Gene Symbols LGALS3BP
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 19-300 of human Galectin-3BP/MAC-2BP/LGALS3BP (NP_005558) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3959.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.