Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody (OTI6B7), PE/Cy5.5, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Galectin-3BP/MAC-2BP/LGALS3BP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI6B7 |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K |
| Gene Symbols | LGALS3BP |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.