Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-4 Antibody (MM0300-1D12), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP212323V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212323V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212323V Supplier Novus Biologicals Supplier No. NBP212323V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin-4 Monoclonal specifically detects Galectin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Galectin-4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0300-1D12
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. P56470
Gene Alias Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4
Gene Symbols LGALS4
Host Species Mouse
Immunogen Human recombinant Galectin-4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3960
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.