Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Galectin 5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$191.80 - $481.00
Specifications
Antigen | Galectin 5 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145326
|
Invitrogen™
PA5145326 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145325
![]() |
Invitrogen™
PA5145325 |
200 μL |
Each for $481.00
|
|
|||||
Description
Galectin 5 is a member of the galectin family of proteins, which are characterized by their ability to bind to specific carbohydrate structures on cell surfaces. Galectin 5 is primarily expressed in the cytoplasm of cells and has been shown to play a role in regulating cell growth, differentiation, and apoptosis. It has also been implicated in various physiological processes, including inflammation, immune response, and cancer progression. Galectin 5 has been found to interact with a variety of proteins and signaling pathways, suggesting that it may have multiple functions within cells.Specifications
Galectin 5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
P47967 | |
25475 | |
Recombinant Rat GAL5 protein, Met1-Thr145 (Accession #P47967). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
Lgals5 | |
Lgals5 | |
IgG | |
Antigen affinity chromatography, Protein A | |
Lgals5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title