Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-7 Antibody, Novus Biologicals™
SDP

Catalog No. NB100380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100380 Supplier Novus Biologicals Supplier No. NB100380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Galectin-7 Polyclonal specifically detects Galectin-7 in Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Galectin-7
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:40000, Immunoprecipitation 10 - 20 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_002298.1
Gene Alias Gal-7, GAL7, galectin 7, galectin-7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1
Gene Symbols LGALS7
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and the C-terminus (residue 135) of human Galectin-7 using the numbering given in entry NP_002298.1 (GeneID 3963).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3963
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.