Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-9 Antibody (1005438), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313840 Supplier Novus Biologicals Supplier No. FAB20455APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Galectin-9 Monoclonal specifically detects Galectin-9 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Galectin-9
Applications Flow Cytometry
Classification Monoclonal
Clone 1005438
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Host Species Mouse
Immunogen E. coli-derived human Galectin-9, Ala2-Thr323, Accession # O00182
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.