Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Galectin-9 Antibody (766428), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB303956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303956 Supplier Novus Biologicals Supplier No. FAB3535V

Rat Monoclonal Antibody

Galectin-9 Monoclonal antibody specifically detects Galectin-9 in Mouse samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen Galectin-9
Applications Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 766428
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ecalectin, Gal-9, galectin 9, galectin-9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Host Species Rat
Immunogen E. coli-derived recombinant mouse Galectin-9 (short isoform), Ala2-Thr322, Accession # O08573-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3965
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.