Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Galeterone
GREENER_CHOICE

Catalog No. p-7121039
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CYP17 inhibitor; also androgen receptor antagonist

Galeterone is a CYP17 inhibitor (IC50 = 300 nM) and androgen receptor antagonist (IC50 = 384 nM in PC3-AR cells). Inhibits DHT-stimulated prostate cancer cell proliferation in vitro. Exhibits antitumor activity in SCID mice bearing prostate cancer xenografts.

Specifications

Chemical Name or Material Galeterone
CAS 851983-85-2
Synonym (3β)-17-(1H-Benzimidazol-1-yl)androsta-5,16-dien-3-ol
Quantity 50 mg
Solubility Soluble to 50mM in DMSO and to 50mM in ethanol
Molecular Formula C26H32N2O
Formula Weight 388.55
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.