Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gallium(III) iodide, ultra dry, 99.999% (metals basis)

Catalog No. AA3569903
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AA3569903

Supplier: Thermo Scientific Chemicals 03569903

Only null left
Add to Cart

CAS: 13450-91-4 | GaI3 | 450.436 g/mol

Gallium(III) iodide is widely used as lewis acid, a precursor of organogallium reagents. It can also be involved in detection of solar neutrinos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-91-4
Molecular Formula GaI3
Molecular Weight (g/mol) 450.436
MDL Number MFCD00016107
InChI Key DWRNSCDYNYYYHT-UHFFFAOYSA-K
Synonym gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basis
PubChem CID 83478
IUPAC Name triiodogallane
SMILES [Ga](I)(I)I

Specifications

Boiling Point 345°C (sublimation)
Melting Point 212°C
Color Yellow to Green
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in aromatic solvents.
Formula Weight 450.43
Sensitivity Light and moisture sensitive
Appearance Yellow-green powder
Density 4.15 g/cm3
Chemical Name or Material Gallium(III) iodide
Show More Show Less
Hazard Category H314-H317-H334-H335
Precautionary Statement P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation DOT Class: 8, Packing Group: II
EINECSNumber 236-611-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.