Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALNT14 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GALNT14 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GALNT14 Polyclonal specifically detects GALNT14 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GALNT14 | |
Polyclonal | |
Rabbit | |
Human | |
EC 2.4.1.41, FLJ12691, FLJ13977, GalNac-T10, GalNac-T14, GALNT15, Polypeptide GalNAc transferase 14, polypeptide N-acetylgalactosaminyltransferase 14, polypeptide N-acetylgalactosaminyltransferase 15, pp-GaNTase 14, Protein-UDP acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T14, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 14 (GalNAc-T14) | |
GALNT14 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
79623 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:IREIILVDDFSNDPDDCKQLIKLPKVKCLRNNERQGLVRSRIRGADIAQGTTLTFLDSHCEVNRDWLQPLLHRVKEDYTRVVCPVIDIINLDTFTYIESASELRGGFDWSLHFQWEQL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title