Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALNT6 Antibody (HL2392) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295142 Supplier Novus Biologicals Supplier No. NBP325475
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

GALNT6 Monoclonal antibody specifically detects GALNT6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen GALNT6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL2392
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.4.1.41, GalNAc transferase 6, GalNAcT6, GALNAC-T6, Polypeptide GalNAc transferase 6, polypeptide N-acetylgalactosaminyltransferase 6, pp-GaNTase 6, Protein-UDP acetylgalactosaminyltransferase 6, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6, UDP-N-acetyl-alpha-D-galactosamine: polypeptideN-acetylgalactosaminyltransferase 6, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 6 (GalNAc-T6)
Host Species Rabbit
Immunogen Recombinant fragment of human GALNT6.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11226
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.