Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Galunisertib

Click to view available options
Quantity:
5 mg
25 mg
Description
Galunisertib is an ALK4 and ALK5 (TGFβRI) inhibitor (IC50 values are 77.7 and 172 nM, respectively). Also inhibits a range of other kinases with submicromolar IC50s including MINK, TGFβRII, ALK6 and ACVR2B. Inhibits TGFβ-induced SMAD phosphorylation in cancer cell lines in vitro and in vivo. Also reverses TGFβ-mediated suppression of T cell proliferation. Prevents TGFβ-mediated EMT and cell migration in vitro and exhibits antitumor efficacy in vivo. Used as component of growth media for urothelial organoids. Orally bioavailable.
Specifications
Specifications
Chemical Name or Material | 4-[5,6-Dihydro-2-(6-methyl-2-pyridinyl)-4 H-pyrrolo[1,2-b]pyrazol-3-yl]-6-quinolinecarboxamide |
CAS | 700874-72-2 |
Quantity | 25 mg |
Target | TGF-beta Receptor Inhibitors |
Molecular Formula | C22H19N5O |
Formula Weight | 369.43 |
Product Type | Inhibitor |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction