Learn More
Description
Specifications
Specifications
| Antigen | gamma Catenin |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | [11E4] |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB |
| Gene Symbols | JUP |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
