Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Antibody (15F11), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP2427A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2427A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2427A32 Supplier Novus Biologicals Supplier No. NBP254527AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen gamma Catenin
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 15F11
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
Gene Symbols JUP
Host Species Mouse
Immunogen Recombinant, full length chicken gamma Catenin protein (Uniprot: P14923)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Cytoskeleton Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3728.0
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Feline
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.