Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Antibody (CTNG/2155R), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB392201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392201 Supplier Novus Biologicals Supplier No. NBP308485AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human, Mouse, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen gamma Catenin
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone CTNG/2155R
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
Host Species Rabbit
Immunogen Recombinant, human full-length gamma Catenin protein (Uniprot: P14923)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3728
Target Species Human, Mouse, Rat, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.