Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gamma-Glutamyltransferase 7 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB335420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB335420 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB335420 Supplier Novus Biologicals Supplier No. NBP298100APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gamma-Glutamyltransferase 7 Polyclonal antibody specifically detects Gamma-Glutamyltransferase 7 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Gamma-Glutamyltransferase 7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias D20S101, dJ18C9.2, gamma-glutamyltransferase 4, gamma-glutamyltransferase 7, gamma-glutamyltransferase-like 3, gamma-glutamyltransferase-like 5, gamma-glutamyltranspeptidase-like 3, GGT4, GGTL5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Gamma-Glutamyltransferase 7 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2686
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.