Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Sarcoglycan Antibody, Novus Biologicals™
SDP

Catalog No. NBP159744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159744 Supplier Novus Biologicals Supplier No. NBP159744
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

gamma Sarcoglycan Polyclonal specifically detects gamma Sarcoglycan in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen gamma Sarcoglycan
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q13326
Gene Alias 35 kDa dystrophin-associated glycoprotein, 35DAG, A4, DAGA4,35kD dystrophin-associated glycoprotein, DMDA, DMDA1, gamma sarcoglycan, gamma-sarcoglycan, Gamma-SG, MAM, MGC130048, sarcoglycan, gamma (35kD dystrophin-associated glycoprotein), sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein), SCARMD2, SCG3LGMD2C, TYPE
Gene Symbols SGCG
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SGCG(sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein)) The peptide sequence was selected from the middle region of SGCG. Peptide sequence FTVDEKEVVVGTDKLRVTGPEGALFEHSVETPLVRADPFQDLRLESPTRS.
Molecular Weight of Antigen 32 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6445
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.