Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Tubulin Antibody, Cy3, Novus Biologicals™
SDP

Catalog No. NB12011319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011319 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011319 Supplier Novus Biologicals Supplier No. NB12011319
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

gamma Tubulin Polyclonal specifically detects gamma Tubulin in Human, Canine, Chicken samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen gamma Tubulin
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.8mg/mL
Conjugate Cyanine3
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation 10 mM PBS (pH 7.4) and 1.0% BSA with 0.05% Sodium Azide
Gene Alias gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide
Gene Symbols TUBG1
Host Species Rabbit
Immunogen Synthetic peptide: EEFATEGTDRKDVFFY-K, corresponding to amino acids 38-53 of Human gamma Tubulin.
Molecular Weight of Antigen 51 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7283
Test Specificity Does not react with alpha, beta, omega and eta Tubulin. The IF testing of this product is performed using chicken fibroblasts. We have not performed IF testing using human cells. This antibody is tested in western blotting using the following cell lysates ; U87, HeLa, Jurkat, C2C12, Rat2, MDCK, and A431.
Target Species Human, Canine, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.